Screw Cap Tubes with Round Base and Colored Screw Caps
Supplier: SARSTEDT INC
60.540.108 60.540.322 60.540.352 60.541.300 60.541.304 60.541.633
77587-716CS 490.57 CAD
77587-716 77587-722 77587-724 77587-732 77587-734 77587-740
Screw Cap Tubes with Round Base and Colored Screw Caps
Tubes Centrifuge Tubes
High-quality, safe and robust containers are essential for mixing, processing and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, Sarstedt offers the right product for every laboratory and analytical requirement.
  • Material: PP
  • Graduations: No
  • Base shape: Round base
  • Corresponds to the requirement: IATA
  • Different cap colors for better coding
  • Polypropylene (PP) tubes for thermal, mechanical and chemical applications
  • Ribbed cap for maximum grip
This broad product portfolio in the field of reagent and centrifuge tubes includes options with round, conical or flat base; with or without skirted base; with stopper or screw cap; with or without print or labels.