Tubes without Cap, Round Base, PS

Supplier: SARSTEDT INC
55.467.006 55.468 62.467.012 55.459 62.476.016 62.476.017 62.476.018 62.476.019 62.476.020 62.476.178 62.476.322 62.476.324 62.476.326 62.492 62.493.300
77587-528CS 219.62 CAD
77587-528 77587-532 77587-858 77587-514 77587-862 77587-864 77587-866 77587-868 77587-870 77587-874 77587-876 77587-878 77587-880 77587-884 77587-886
Tubes without Cap, Round Base, PS
Tubes Centrifuge Tubes
High-quality, safe and robust containers are essential for mixing, processing and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, this range offers the right product for every laboratory and analytical requirement.

  • Material: PS
  • Base shape: Round base
  • Amount of packaging reduced by 40% (if StackPack option is used)
  • Polystyrene (PS) tubes for maximum transparency
  • Different push caps for a variety of applications
  • Volumes from 3.5 to 26 ml

This broad product portfolio in the field of reagent and centrifugeMore Product Information
New Product
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR