Blood Gas Monovette®, 1 and 2 ml
Supplier: SARSTEDT INC
05.1147.028 05.1147.048
77587-288CS 877.58 CAD
77587-288 77587-290
Blood Gas Monovette®, 1 and 2 ml
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
The blood gas Monovette® minimizes diagnostic blood loss in patients that require special monitoring. In addition, the fluid-dosed Ca²⁺-balanced heparin ensures the rapid and optimal mixing of blood and anticoagulant.
  • Preparation: Calcium-balanced lithium heparin
  • For arterial and venous blood gas analysis
  • For whole blood extraction (LH)
  • Preparation: Calcium-balanced lithium heparin in droplet form as a spray dose; 50 IU heparin/ml blood
  • Calcium (Ca²⁺) balanced heparin is also suitable for determining electrolytes
The blood gas Monovette® is available for collecting specimens of arterial and/or venous blood and can be used to determine blood gas, pH or electrolytes, for example, on a blood gas analyzer. As the anticoagulant is present in droplet form (calcium-balanced heparin), this ensures that it quickly and properly mixes with the blood.