Prepared Sample Tubes with Push Caps and Round Bottom
Supplier: SARSTEDT INC
32.329.100 36.336.100
77587-452CS 516.06 CAD
77587-452 77587-454
Prepared Sample Tubes with Push Caps and Round Bottom
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
These quality sample tubes are prepared with standard additives for individual blood collection requirements.
  • Material: PP
  • Base: Round base
  • Sample tubes for a range of volumes
  • Standard preparations available
  • Ideal for a wide variety of requirements
  • With graduations to check fill level
SARSTEDT offers a range of specimen collection products for optimal sample quality and user safety.
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable