Monovette® Urine Tubes
Supplier: SARSTEDT INC
10.252.028 10.250.001
77587-328CS 250.62 CAD
77587-328 77587-320
Monovette® Urine Tubes
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
In order to meet the various requirements of hygienic collection, safe transportation and storage of diagnostic sample materials, this range offers many products for collecting urine, stool, saliva, CSF and tissue samples.
  • Material: PP
  • Preparation: Without preparation
  • Closure type: Screw cap
  • Light protection: No
  • Hygiene and protection for patients and users
  • Completely needle-free sample collection and processing
  • Entire routine diagnostics can be performed easily from one container
  • Primary container for transport as per ADR and IATA
  • With graduations to check the filling level
The urine Monovette® enables hygienic, needle-free sample collection from urine containers and urine drainage systems. Subsequently, a urine analysis with commercially available test strips can be carried out directly in the filled urine Monovette®. If necessary, the urine Monovette® is also suitable for collecting sediment in centrifuges and for further urine analysis in fully automated urine analysis systems. The Urine Monovette® can also be used as a transport container as per P650 of the ADR and IATA.