Safety-Heel® Incision Lancets
Supplier: SARSTEDT INC
85.1025 85.1026
77589-428CS 1067.57 CAD
77589-428 77589-430
Safety-Heel® Incision Lancets
Blood Lancets
The safety lancets guarantee optimal safety for patients and users, and are flexible with regard to penetration depth and required quantity of blood. The needle and blade are always safely concealed in the lancet body before and after use, which prevents needlestick injuries and cross-contamination.
  • Heel punctures are used to examine premature and newborn babies for life-threatening, congenital diseases
  • Simple operation
  • Optimal blood flow without additional rubbing or massaging with a low penetration depth
  • The crescent-shaped incision path can help to minimize the formation of hematomas
  • Quick healing due to low penetration depth
The pre-analytical product range includes products for optimal specimen collection. The capillary collection systems are among the most versatile on the market.