NANOCOLOR® Photometric Tube Tests, Ammonium
Supplier: CTL SCIENTIFIC SUPPLY CORP
985002 985003 985004 985005 985006 985008
77578-422EA 104.53 CAD
77578-422 77578-424 77578-416 77578-426 77578-420 77578-418
NANOCOLOR® Photometric Tube Tests, Ammonium
Environmental Test Kits
A range of analytical reagents for photometric determination of ammonium in water analysis using a pre-dosed tube.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Bar code cuvette identification
  • Precisely pre-dosed reagents in 16 mm (Ext.Ø) tubes
Tests that are particularly suitable for the NANOCOLOR® photometers but can be used with other photometers.