S-Monovette® Safety and Non Safety Needles
Supplier: SARSTEDT INC
85.1372 85.1441 85.1160
77589-442CS 546.33 CAD
77589-442 77589-454 77589-432
S-Monovette® Safety and Non Safety Needles
Syringe Needles
The high quality of blood samples collected with the S-Monovette® blood collection system is due to the gentle aspiration technique. It has been proven to significantly reduce hemolysis rates compared to vacuum systems. As a result, erythrocytes remain intact and blood collection does not have to be repeated as often.
  • Safety needle come with integrated holder
  • Ready to use immediately - no complicated assembly of needle and holder required
  • Flat puncturing angle ensures safe puncture
  • Needle guard can be activated with one hand for maximum handling convenience; safety needle guard complies with national and international guidelines such as TRBA250, EU Directive 2010/32/EU
  • Sterile, non-pyrogenic/endotoxin-free
Due to the gentle aspiration technique, the S-Monovette® blood collection system is suitable for all vein conditions, from young to old. It offers optimal sample quality and the best results thanks to the revolutionary blood collection system.
This also includes a well-designed, environmentally friendly packaging concept to reduce waste and a ready-to-use system for our needles. All needles can be used straight out of the packaging and allow users to work quickly - there is no assembly required as the needles are available exclusively with integrated holders.
In addition to a premium blood collection system, we support our customers in their everyday work with a pre-analytical consulting group. A global team of experts is available for any pre-analytical questions, areas needing improvement, or product training.