Crystal Digital PCR® Assay for C. albicans
Supplier: STILLA TECHNOLOGIES, INC.
R52002.1 R52002.2
77590-202EA 5194.66 CAD
77590-202 77590-204
Crystal Digital PCR® Assay for C. albicans
Assays qPCR Assays
Assay-specific Crystal Flex Probes and primers for the detection of C. albicans and its Positive Control. C. albicans Crystal Digital PCR® Assay is a 10X assay designed to detect and quantify 16 genes (1 gene per chromosome) of Candida albicans using the Ruby Chip. Candida albicans is a common yeast within the human gastrointestinal microbiota. Under certain conditions, it can become pathogenic leading to infections.
  • Ready-to-use assay reagents to detect multiple targets in a single reaction
  • Crystal Flex Probe detection chemistry for flexible assay configuration
  • Same set of Crystal Universal Reporters to detect any target in different assays
  • High multiplexing with low price per point
Wet lab tested
Compatibility: Nio™ Digital PCR, Prism6, ruby chip
Channels: Blue, red, teal, green, yellow, infra-red
Required reagents: Crystal universal reporter 7 and naica® PCR MIX 10X
Target: C. albicans (1 gene for each of 16 chromosomes)
Caution: For research use only. Not for use in diagnostic procedures.