Crystal Digital PCR® Assay for HPV (ALB, HPV16/18/31/33/35/39/45/51/52/56/58)
Supplier: STILLA TECHNOLOGIES, INC.
Assay-specific Crystal Flex Probes and primers for the detection of HPV genotypes and its Positive Control. HPV (ALB, HPV16/18/31/33/35/39/45/51/52/56/58) Crystal Digital PCR® Assay is a 10X assay designed to detect and quantify 11 high risk HPV genotypes for cervical cancer plus a reference gene for human DNA quantification using the Ruby Chip.
- Ready-to-use assay reagents to detect multiple targets in a single reaction
- Crystal Flex Probe detection chemistry for flexible assay configuration
- Same set of Crystal Universal Reporters to detect any target in different assays
- High multiplexing with low price per point
Wet lab tested
Compatibility: Nio™ Digital PCR, Prism6, ruby chip
Channels: Blue, red, teal, green, yellow, infra-red
Required reagents: Crystal universal reporter 7 and naica® PCR MIX 10X
Target: ALB, HPV16, HPV18, HPV31, HPV33, HPV35, HPV39, HPV45, HPV51, HPV52, HPV56, HPV58
Caution: For research use only. Not for use in diagnostic procedures.