Nitrile Powder Free Examination Gloves, Chemo Rated
Supplier: Omni International
212-21 212-22 212-23 212-24
77587-142BX 12.83 CAD
77587-142 77587-144 77587-146 77587-148
Nitrile Powder Free Examination Gloves, Chemo Rated
Gloves
OmniTrust 212 series nitrile powder free examination gloves, chemo rated is a thermally activated glove that starts conforming to your hand when donned.
  • Ambidextrous beaded cuff, single use only
  • Comfortable formulation, thermally activated
  • Tested for safe use with chemo drugs
  • Not made with natural rubber latex
  • Textured fingers for tactile sensitivity
  • Fentanyl resistant
The comfort formulation, reducing hand fatigue. It has micro-texture fingertips for added grip control without loss of tactile sensitivity. This series is also tested safe use with chemotherapy drugs.