Crystal Digital PCR® Assay for BRCA1 Methylation
Supplier: STILLA TECHNOLOGIES, INC.
R51034.1 R51034.2
77590-190EA 389.61 CAD
77590-190 77590-192
Crystal Digital PCR® Assay for BRCA1 Methylation
Assays qPCR Assays
Assay-specific Crystal Flex Probes and primers for the detection of BRCA1 Methylation and its Positive Control. The BRCA1 Methylation Crystal Digital PCR® Assay is a 10X assay designed to detect and quantify methylation of the BRCA1 promoter using the Ruby Chip. BRCA1 is crucial for maintaining genomic stability by repairing DNA damage, regulating the cell cycle, and acting as a tumor suppressor to prevent cancer.
  • Ready-to-use assay reagents to detect multiple targets in a single reaction
  • Crystal Flex Probe detection chemistry for flexible assay configuration
  • Same set of Crystal Universal Reporters to detect any target in different assays
Wet lab tested
Compatibility: Nio™ Digital PCR, Prism6, ruby chip
Channels: Yellow, red, infra-red
Required reagents: Crystal universal reporter 7 and naica® PCR MIX 10X
Target: BRCA1 promoter methylation
Caution: For research use only. Not for use in diagnostic procedures.