Crystal Digital PCR® Assay for NRAS (G12D, drop-off G12-G13)
Supplier: STILLA TECHNOLOGIES, INC.
Assay-specific Crystal Flex Probes and primers for the detection of NRAS mutations and its positive control. NRAS (G12D, drop-off G12-G13) Crystal Digital PCR® Assay is a 10X assay designed to detect and quantify mutations in G12-G13 plus 1 mutation of the NRAS gene using the Ruby Chip. NRAS is pivotal in regulating cell signaling pathways implicated in cancer development, notably melanoma and colorectal cancer.
- Ready-to-use assay reagents to detect multiple targets in a single reaction
- Crystal Flex Probe detection chemistry for flexible assay configuration
- Same set of Crystal Universal Reporters to detect any target in different assays
Wet lab tested
Compatibility: Nio™ Digital PCR, Prism6, Prism3, ruby chip
Channels: Blue, green, red
Required reagents: Crystal universal reporter 3 and naica® PCR MIX 10X
Target: NRAS G12-G13/G12D
Caution: For research use only. Not for use in diagnostic procedures.