NANOCOLOR® Photometric Tube Tests, Formaldehyde
Supplier: CTL SCIENTIFIC SUPPLY CORP
985041 985046
77578-500EA 123.84 CAD
77578-500 77578-498
NANOCOLOR® Photometric Tube Tests, Formaldehyde
Environmental Test Kits
A range of pre-dosed NANOCOLOR® tube tests for photometric determination of formaldehyde in water.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Barcode cuvette identification
  • Precisely pre-dosed reagents in ext.Ø 16 mm tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.