NANOCOLOR® Photometric Tube Tests, Anionic Surfactants
Supplier: Macherey-Nagel
NANOCOLOR®
985032
77578-428EA 126.94 CAD
77578-428
NANOCOLOR® Photometric Tube Tests, Anionic Surfactants
Environmental Test Kits
A pre-dosed NANOCOLOR tube test for photometric determination of anionic surfactants in water.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Barcode cuvette identification
  • Precisely pre-dosed reagents in ext.Ø 16 mm tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.