Crystal Digital PCR® Assay for TP53 (drop-off V272-A276/R283-E286)
Supplier: STILLA TECHNOLOGIES, INC.
R51030.1 R51030.2
77590-174EA 389.61 CAD
77590-174 77590-176
Crystal Digital PCR® Assay for TP53 (drop-off V272-A276/R283-E286)
Assays qPCR Assays
Assay-specific Crystal Flex Probes and primers for the detection of TP53 mutations and its positive control. TP53 (drop-off V272-A276/R283-E286) Crystal Digital PCR® Assay is a 10X assay designed to detect and quantify mutations between V272-A276 or R283-E286 of the TP53 gene using the Ruby Chip. TP53 is pivotal in regulating cell divisison, thus an important tumor suppressor gene.
  • Ready-to-use assay reagents to detect multiple targets in a single reaction
  • Crystal Flex Probe detection chemistry for flexible assay configuration
  • Same set of Crystal Universal Reporters to detect any target in different assays
Wet lab tested
Compatibility: Nio™ Digital PCR, Prism6, Prism3, ruby chip
Channels: Blue, green
Required reagents: Crystal universal reporter 3 and naica® PCR MIX 10X
Target: TP53 V272-A276 and R283-E286
Caution: For research use only. Not for use in diagnostic procedures.