BoneClones® Economy Human Female African-American Comparative Set
Supplier: BONE CLONES, INC.
BoneClones®
COMP-112
470356-974KT 945.16 CAD
470356-974
BoneClones® Economy Human Female African-American Comparative Set
Models Human Biology Models
This affordable set was created to introduce the student to the human skeleton. The skull and pelvis are elements that can be used to discuss sexual dimorphism and ancestral affiliation, as well as concepts of human evolution and development.
Additionally, the humerus and femur can be used to examine aging processes and stature estimation. This set is an excellent addition to any introductory program going over forensic science, human evolution, biological anthropology, and human biology.
Delivery information: Contains human female african-american skull, pelvis, femur and humerus.