Crystal Digital PCR® Assay for NRAS (Q61, Q61K)
Supplier: STILLA TECHNOLOGIES, INC.
R51027.1 R51027.2
77590-162EA 389.61 CAD
77590-162 77590-164
Crystal Digital PCR® Assay for NRAS (Q61, Q61K)
Assays qPCR Assays
Assay-specific Crystal Flex Probes and primers for the detection of NRAS mutations and its Positive Control. NRAS (Q61, Q61K) Crystal Digital PCR® Assay is a 10X assay designed to detect and quantify 1 mutation in the NRAS gene using the Ruby Chip. NRAS is pivotal in regulating cell signaling pathways implicated in cancer development, notably melanoma and colorectal cancer.
  • Ready-to-use assay reagents to detect multiple targets in a single reaction
  • Crystal Flex Probe detection chemistry for flexible assay configuration
  • Same set of crystal universal reporters to detect any target in different assays
Wet lab tested
Compatibility: Nio™ Digital PCR, Prism6, Prism3, ruby chip
Channels: Blue, red
Required reagents: Crystal universal reporter 3 and naica® PCR MIX 10X
Target: NRAS Q61/Q61K
Caution: For research use only. Not for use in diagnostic procedures.