Crystal Digital PCR® Assay for EGFR (drop-off Del19)
Supplier: STILLA TECHNOLOGIES, INC.
R51026.1 R51026.2
77590-158EA 389.61 CAD
77590-158 77590-160
Crystal Digital PCR® Assay for EGFR (drop-off Del19)
Assays qPCR Assays
Assay-specific Crystal Flex Probes and primers for the detection of EGFR mutations and its Positive Control. The EGFR (drop-off Del19) Crystal Digital PCR® Assay is a 10X assay designed to detect and quantify EGFR Exon 19 deletion mutations using the Ruby Chip. EGFR encodes for the epidermal growth factor receptor, a protein that plays a crucial role in cell growth, proliferation, and differentiation.
  • Ready-to-use assay reagents to detect multiple targets in a single reaction
  • Crystal Flex Probe detection chemistry for flexible assay configuration
  • Same set of crystal universal reporters to detect any target in different assays
Wet lab tested
Compatibility: Nio™ Digital PCR, Prism6, Prism3, ruby chip
Channels: Blue, green
Required reagents: Crystal universal reporter 3 and naica® PCR MIX 10X
Target: EGFR Deletion exon 19
Caution: For research use only. Not for use in diagnostic procedures.