Crystal Digital PCR® Assay for EGFR (L858, L858R, L861Q)
Supplier: STILLA TECHNOLOGIES, INC.
R51025.1 R51025.2
77590-154EA 519.47 CAD
77590-154 77590-156
Crystal Digital PCR® Assay for EGFR (L858, L858R, L861Q)
Assays qPCR Assays
Assay-specific Crystal Flex Probes and primers for the detection of EGFR mutations and its Positive Control. The EGFR (L858, L858R, L861Q) Crystal Digital PCR® Assay is a 10X assay designed to detect and quantify 2 mutations in the EGFR gene using the Ruby Chip. EGFR encodes for the epidermal growth factor receptor, a protein that plays a crucial role in cell growth, proliferation, and differentiation.
  • Ready-to-use assay reagents to detect multiple targets in a single reaction
  • Crystal Flex Probe detection chemistry for flexible assay configuration
  • Same set of crystal universal reporters to detect any target in different assays
Wet lab tested
Compatibility: Nio™ Digital PCR, Prism6, Prism3, ruby chip
Channels: Blue, green, red
Required reagents: Crystal universal reporter 3 and naica® PCR MIX 10X
Target: EGFR L858/L858R/L861Q
Caution: For research use only. Not for use in diagnostic procedures.