Polypropylene Tubes with Round Base
Supplier: SARSTEDT INC
60.557.010 60.557.011 55.535.300 55.539 55.598.004 55.598.006 62.526.003 55.516.300 55.522.300 55.525.061 55.516.016 55.526.002 55.526.045 55.526.046 55.526.521 55.526.522 55.526.523 55.526.524 55.526.525 55.526.526 55.526.531 55.526.532 55.526.533 55.526.534 55.526.535 55.532.300
77587-782CS 344.6 CAD
77587-782 77587-784 77587-644 77587-648 77587-650 77587-652 77587-904 77587-584 77587-590 77587-600 77587-580 77587-604 77587-608 77587-610 77587-616 77587-618 77587-620 77587-622 77587-624 77587-626 77587-628 77587-630 77587-632 77587-634 77587-636 77587-640
Polypropylene Tubes with Round Base
Tubes Centrifuge Tubes
High-quality, safe and robust containers are essential for mixing, processing and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
  • Material: Polypropylene (PP)
  • Amount of packaging reduced by 40% (if StackPack option is used)
  • Polypropylene (PP) tubes for thermal, mechanical and chemical applications
  • Different push caps for a variety of applications
  • Volumes from 1 to 30 ml
This broad product portfolio in the field of reagent and centrifuge tubes includes options with round, conical or flat base; with or without skirted base; with stopper or screw cap; with or without print or labels.