Crystal Digital PCR® Assay for NRAS (drop-off Q61)
Supplier: STILLA TECHNOLOGIES, INC.
Assay-specific Crystal Flex Probes and primers for the detection of NRAS mutations and its positive control.The NRAS (drop-off Q61) Crystal Digital PCR® Assay is a 10X assay designed to detect and quantify mutations in Q61 of the NRAS gene using the Ruby Chip. NRAS is pivotal in regulating cell signaling pathways implicated in cancer development, notably melanoma and colorectal cancer.
- Ready-to-use assay reagents to detect multiple targets in a single reaction
- Crystal Flex Probe detection chemistry for flexible assay configuration
- Same set of crystal universal reporters to detect any target in different assays
Wet lab tested
Compatibility: Nio™ Digital PCR, Prism6, Prism3, ruby chip
Channels: Green, blue
Required reagents: Crystal universal reporter 3 and naica® PCR MIX 10X
Target: NRAS Q61
Caution: For research use only. Not for use in diagnostic procedures.