Crystal Digital PCR® Assay for MYD88 (L265, L265P)
Supplier: STILLA TECHNOLOGIES, INC.
R51015.1 R51015.2
77590-114EA 389.61 CAD
77590-114 77590-116
Crystal Digital PCR® Assay for MYD88 (L265, L265P)
Assays qPCR Assays
Assay-specific Crystal Flex Probes and primers for the detection of MYD88 mutations and its positive control. The MYD88 (L265, L265P) Crystal Digital PCR® Assay is a 10X assay designed to detect and quantify 1 mutation in the MYD88 gene using the Ruby Chip. MYD88 encodes the MYD88 protein, which is an adaptor protein essential for transmitting signals in the immune system, mediating responses to infections, and modulating inflammation and cell survival.
  • Ready-to-use assay reagents to detect multiple targets in a single reaction
  • Crystal Flex Probe detection chemistry for flexible assay configuration
  • Same set of crystal universal reporters to detect any target in different assays
Wet lab tested
Compatibility: Nio™ Digital PCR, Prism6, Prism3, ruby chip
Channels: Red, blue
Required reagents: Crystal universal reporter 3 and naica® PCR MIX 10X
Target: MYD88 L265, L265P
Caution: For research use only. Not for use in diagnostic procedures.