SteriPlast® Detectable Spoons, Long Handle, Blue
Supplier: BURKLE INC
5378-5004 5378-5008
77581-256PK 69.58 CAD
77581-256 77581-258
SteriPlast® Detectable Spoons, Long Handle, Blue
Spoons
The sampling spoon with long handle can be used to remove powders, granulates, pastes or liquids. The long handle makes it easy to reach the bottoms of larger containers as well and allows for sampling of drums, jerrycans and bottles with small opening diameters. Thanks to the indentations on the handle, the disposable spoon rests comfortably and securely in the hand.
  • PS detectable
  • Individually packaged and sterilized, for single use
  • Can be detected by color scanners, metal detectors or X-rays
  • Can be used as part of HACCP/IFS/BRC foreign object management
  • Handle length 300 mm
  • Clean room manufactured (clean room class seven)
The laboratory spoon is suitable for single use in the sensitive areas of pharmaceuticals, chemistry, cosmetics, animal feed and food and beverage.
Certifications: Complies with EU food requirements and FDA regulations.