SteriPlast® Detectable Spoons
Supplier: BURKLE INC
5378-2011 5378-2012
77581-224PK 134.19 CAD
77581-224 77581-226
SteriPlast® Detectable Spoons
Spoons
These detectable SteriPlast® spoons, made of PS, are produced in a class seven cleanroom (10000), individually packaged for disposable use and then sterilised by gamma rays. The detectable spoons are available in two sizes, 2.5 ml (similar to a teaspoon) and 10 ml (similar to a tablespoon).
  • Can be detected by color scanners, metal detectors or X-rays
  • Individually packaged and sterilized by gamma rays
  • Production, assembly and packaging according to cleanroom class seven
These spoons are not only colored blue but also have a special additive in the material that makes them visible to metal detectors or X rays. These products can therefore be rejected with standard systems for checking for foreign objects, even though they are made of plastic.
Certifications: Complies with EU food requirements and FDA regulations.