SteriPlast® Detectable Scoops, Long Handle, Blue
Supplier: BURKLE INC
5378-5016 5378-5020
77581-262PK 91.67 CAD
77581-262 77581-264
SteriPlast® Detectable Scoops, Long Handle, Blue
Scoops
The sampling scoop can be used to take samples of powders, granulates and pastes. The long handle makes it possible to sample larger containers even with little content or to reach through narrow openings. With the straight bottom of the scoop, the medium can be scooped up directly from the bottom of the container.
  • PS detectable
  • Individually packaged and sterilized, for single use
  • Can be detected by color scanners, metal detectors or X-rays
  • Can be used as part of HACCP/IFS/BRC foreign object management
  • Handle length 300 mm
  • Clean room manufactured (clean room class seven)
The disposable scoop is suitable for sensitive areas of pharmaceuticals and chemistry as well as the food and cosmetics industries.
Certifications: Complies with EU food requirements and FDA regulations.