NANOCOLOR® Photometric Tube Tests, Nitrite
Supplier: CTL SCIENTIFIC SUPPLY CORP
985068 985069
77578-526EA 62.81 CAD
77578-526 77578-528
NANOCOLOR® Photometric Tube Tests, Nitrite
Environmental Test Kits
A pre-dosed NANOCOLOR® tube test for photometric determination of nitrite (NO₂-) in water.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Barcode cuvette identification
  • Precisely pre-dosed reagents in 16 mm (OD) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.