NANOCOLOR® Photometric Tube Tests, Nitrate
Supplier: CTL SCIENTIFIC SUPPLY CORP
985064 985065 985066
77578-522EA 83.74 CAD
77578-522 77578-524 77578-520
NANOCOLOR® Photometric Tube Tests, Nitrate
Environmental Test Kits
A pre-dosed NANOCOLOR® tube test for photometric determination of nitrate in water.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Barcode cuvette identification
  • Precisely pre-dosed reagents in 16 mm (OD) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.