NANOCOLOR® Photometric Tube Tests, Sulphate
Supplier: CTL SCIENTIFIC SUPPLY CORP
985063 985060 985062
77578-566EA 137.67 CAD
77578-566 77578-570 77578-568
NANOCOLOR® Photometric Tube Tests, Sulphate
Environmental Test Kits
A pre-dosed NANOCOLOR® tube test for photometric determination of sulphate in water.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Barcode cuvette identification
  • Precisely pre-dosed reagents in 16 mm (OD) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.