NANOCOLOR® Photometric Tube Tests, Zinc
Supplier: CTL SCIENTIFIC SUPPLY CORP
985042
985096
77578-602EA
206.11
CAD
77578-602
77578-600
NANOCOLOR® Photometric Tube Tests, Zinc
Environmental Test Kits
A pre-dosed NANOCOLOR® tube test for photometric determination of zinc in water.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Barcode cuvette identification
- Precisely pre-dosed reagents in 16 mm (OD) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.