NANOCOLOR® Photometric Tube Tests, Sulphite
Supplier: CTL SCIENTIFIC SUPPLY CORP
985089
985090
77578-574EA
194.85
CAD
77578-574
77578-576
NANOCOLOR® Photometric Tube Tests, Sulphite
Environmental Test Kits
A pre-dosed NANOCOLOR® tube test for photometric determination of sulphite in water.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Barcode cuvette identification
- Precisely pre-dosed reagents in 16 mm (OD) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.