NANOCOLOR® Photometric Tube Tests, Total Nitrogen
Supplier: CTL SCIENTIFIC SUPPLY CORP
985088 985092 985083
77578-590EA 144.76 CAD
77578-590 77578-594 77578-592
NANOCOLOR® Photometric Tube Tests, Total Nitrogen
Environmental Test Kits
A pre-dosed NANOCOLOR® tube test for photometric determination of total nitrogen in water.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Barcode cuvette identification
  • Precisely pre-dosed reagents in 16 mm (ext.Ø) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.