NANOCOLOR® Photometric Tube Tests, Copper
Supplier: CTL SCIENTIFIC SUPPLY CORP
985053 985054
77578-484EA 139.45 CAD
77578-484 77578-486
NANOCOLOR® Photometric Tube Tests, Copper
Environmental Test Kits
A range of pre-dosed NANOCOLOR® tube tests for photometric determination of the copper in water.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Barcode cuvette identification
  • Precisely pre-dosed reagents in 16 mm (OD) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.