NANOCOLOR® Photometric Tube Tests, Starch
Supplier: Macherey-Nagel
NANOCOLOR®
985085
77578-564EA 172.5 CAD
77578-564
NANOCOLOR® Photometric Tube Tests, Starch
Environmental Test Kits
A pre-dosed NANOCOLOR® tube test for photometric determination of starch in water.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Barcode cuvette identification
  • Precisely pre-dosed reagents in 16 mm (OD) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.