NANOCOLOR® Photometric Tube Tests, ortho- and Total Phosphate
Supplier: CTL SCIENTIFIC SUPPLY CORP
985095
985076
985079
985080
985081
985055
77578-536EA
67.31
CAD
77578-536
77578-538
77578-544
77578-540
77578-546
77578-542
NANOCOLOR® Photometric Tube Tests, ortho- and Total Phosphate
Environmental Test Kits
A pre-dosed NANOCOLOR® tube test for photometric determination of ortho- and total phosphate in water.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Barcode cuvette identification
- Precisely pre-dosed reagents in 16 mm (ext.Ø) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.