NANOCOLOR® Photometric Tube Tests, TOC
Supplier: CTL SCIENTIFIC SUPPLY CORP
985075 985078
77578-584EA 288.24 CAD
77578-584 77578-586
NANOCOLOR® Photometric Tube Tests, TOC
Environmental Test Kits
A pre-dosed NANOCOLOR® tube test for photometric determination of TOC (total organic carbon) in water.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Barcode cuvette identification
  • Precisely pre-dosed reagents in 16 mm (ext.Ø) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.