Thermal Transfer Slotted Cassettes, PiSmart
Supplier: StatLab
CAS-PI100SL-WH
CAS-PI100SL-GR
CAS-PI100SL-GY
CAS-PI100SL-BL
CAS-PI100SL-OR
CAS-PI100SL-PK
CAS-PI100SL-YL
CAS-PI100SL-TN
CAS-PI100SL-LV
77575-600CS
1134.47
CAD
77575-600
77575-602
77575-604
77575-606
77575-608
77575-610
77575-612
77575-614
77575-616
Thermal Transfer Slotted Cassettes, PiSmart
Embedding Cassettes
Tissue Processing Cassettes
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...