Crystal Digital PCR® Assay for c-KIT (D816-D816V)
Supplier: STILLA TECHNOLOGIES, INC.
R51010.1 R51010.2
77590-094EA 389.61 CAD
77590-094 77590-096
Crystal Digital PCR® Assay for c-KIT (D816-D816V)
Assays qPCR Assays
Assay-specific crystal flex probes and primers for the detection of c-KIT mutations and its positive control. The c-KIT (D816, D816V) Crystal Digital PCR® Assay is a 10X assay designed to detect and quantify 1 mutation in the c-KIT gene using the Ruby Chip. c-KIT encodes for a receptor tyrosine kinase that plays a crucial role in cell signaling for processes including cell survival, proliferation, and differentiation.
  • Ready-to-use assay reagents to detect multiple targets in a single reaction
  • Crystal flex probe detection chemistry for flexible assay configuration
  • Same set of crystal universal reporters to detect any target in different assays
Wet lab tested
Compatibility: Nio™ Digital PCR, Prism6, Prism3, ruby chip
Channels: Blue, red
Required reagents: Crystal universal reporter 3 and naica® PCR MIX 10X
Targets: c-KIT D816/D816V
Caution: For research use only. Not for use in diagnostic procedures.