Crystal Digital PCR® Assay for ESR1 (11 Mutations)
Supplier: STILLA TECHNOLOGIES, INC.
R51009.1 R51009.2
77590-090EA 3792.11 CAD
77590-090 77590-092
Crystal Digital PCR® Assay for ESR1 (11 Mutations)
Assays qPCR Assays
Assay-specific crystal flex probes and primers for the detection of ESRI mutations and its positive control. ESR1 (11 mutations) Crystal Digital PCR® assay is a 10X assay designed to detect and quantify 11 mutations in the ESR1 gene using the ruby chip. ESR1 is pivotal in mediating resistance to endocrine therapy in metastatic hormone-positive breast cancer.
  • Ready-to-use assay reagents to detect multiple targets in a single reaction
  • Crystal flex probe detection chemistry for flexible assay configuration
  • Same set of crystal universal reporters to detect any target in different assays
Wet lab tested
Compatibility: Nio™ Digital PCR, Prism6, ruby chip
Channels: Blue, teal, green, yellow, red, infra-red
Required reagents: Crystal universal reporter 7 and naica® PCR MIX 10X
Targets: ESR1 E380Q/V422del/S463P/D538G/L536H/L536P/L536R/Y537C/Y537D/Y537N/Y537S
Caution: For research use only. Not for use in diagnostic procedures.