Crystal Digital PCR® Assay for KRAS (G12A, G12C, G12D, G12V, drop-off G12-G13)
Supplier: STILLA TECHNOLOGIES, INC.
R51007.1 R51007.2
77590-082EA 909.07 CAD
77590-082 77590-084
Crystal Digital PCR® Assay for KRAS (G12A, G12C, G12D, G12V, drop-off G12-G13)
Assays qPCR Assays
Assay-specific crystal flex probes and primers for the detection of KRAS mutations and its positive control. The KRAS (G12A, G12C, G12D, G12V, drop-off G12-G13) Crystal Digital PCR® assay is a 10X assay designed to detect and quantify mutations in codons 12 and 13 plus 4 mutations in the KRAS gene using the ruby chip. KRAS is pivotal in regulating cell signaling pathways implicated in cancer development, notably melanoma and colorectal cancer.
  • Ready-to-use assay reagents to detect multiple targets in a single reaction
  • Crystal flex probe detection chemistry for flexible assay configuration
  • Same set of crystal universal reporters to detect any target in different assays
Wet lab tested
Compatibility: Nio™ Digital PCR, Prism6, ruby chip
Channels: Blue, teal, green, yellow, red, infra-red
Required reagents: Crystal universal reporter 7 and naica® PCR MIX 10X
Targets: KRAS G12-G13/G12A/G12C/G12D/G12V
Caution: For research use only. Not for use in diagnostic procedures.