Crystal Digital PCR® Assay for RAD51C Methylation
Supplier: STILLA TECHNOLOGIES, INC.
R51005.1 R51005.2
77590-074EA 519.47 CAD
77590-074 77590-076
Crystal Digital PCR® Assay for RAD51C Methylation
Assays qPCR Assays
Assay-specific crystal flex probes and primers for the detection of RAD51C Methylation and its positive control. The RAD51C methylation Crystal Digital PCR® assay is a 10X assay designed to detect and quantify methylation of the RAD51C promoter using the ruby chip. RAD51C gene is involved in DNA repair, particularly in the homologous recombination pathway for genome stability.
  • Ready-to-use assay reagents to detect multiple targets in a single reaction
  • Crystal flex probe detection chemistry for flexible assay configuration
  • Same set of crystal universal reporters to detect any target in different assays
Wet lab tested
Compatibility: Nio™ Digital PCR, Prism6, ruby chip
Channels: Green, red, infra-red
Required reagents: Crystal universal reporter 7 and naica® PCR MIX 10X
Targets: RAD51C promoter methylation
Caution: For research use only. Not for use in diagnostic procedures.