Crystal Digital PCR® Assay for GSTP1 Methylation
Supplier: STILLA TECHNOLOGIES, INC.
R51003.1 R51003.2
77590-066EA 519.47 CAD
77590-066 77590-068
Crystal Digital PCR® Assay for GSTP1 Methylation
Assays qPCR Assays
Assay-specific crystal flex probes and primers for the detection of GSTP1 methylation and its positive control. The GSTP1 methylation Crystal Digital PCR® assay is a 10X assay designed to detect and quantify methylation of the GSTP1 promoter using the ruby chip. The GSTP1 promoter is implicated on the GSTP1 epigenetic silencing and dysregulation of detoxification pathways contributing to tumor initiation, progression, and drug resistance.
  • Ready-to-use assay reagents to detect multiple targets in a single reaction
  • Crystal flex probe detection chemistry for flexible assay configuration
  • Same set of crystal universal reporters to detect any target in different assays
Wet lab tested
Compatibility: Nio™ Digital PCR, Prism6, ruby chip
Channels: Green, yellow, red
Required reagents: Crystal universal reporter 7 and naica® PCR MIX 10X
Targets: GSTP1 promoter methylation
Caution: For research use only. Not for use in diagnostic procedures.