Crystal Digital PCR® Assay for KRAS (G12, G12C)
Supplier: STILLA TECHNOLOGIES, INC.
Assay-specific crystal flex probes and primers for the detection of KRAS mutations and its positive control. The KRAS (G12, G12C) Crystal Digital PCR® assay is a 10X assay designed to detect and quantify one mutation in the KRAS gene using the ruby chip. KRAS is pivotal in regulating cell signaling pathways implicated in cancer development, notably melanoma and colorectal cancer.
- Ready-to-use assay reagents to detect multiple targets in a single reaction
- Crystal flex probe detection chemistry for flexible assay configuration
- Same set of crystal universal reporters to detect any target in different assays
Wet lab tested
Compatibility: Nio™ Digital PCR, Prism6, Prism3, ruby chip
Channels: Blue, red
Required reagents: Crystal universal reporter 3 and naica® PCR MIX 10X
Targets: KRAS G12/G12C
Caution: For research use only. Not for use in diagnostic procedures.