Crystal Digital PCR® Assay for KRAS (drop-off G12-G13)
Supplier: STILLA TECHNOLOGIES, INC.
R51000.1 R51000.2
77590-054EA 389.61 CAD
77590-054 77590-056
Crystal Digital PCR® Assay for KRAS (drop-off G12-G13)
Assays qPCR Assays
Assay-specific crystal flex probes and primers for the detection of KRAS mutations and its positive control. The KRAS (drop-off G12-G13) Crystal Digital PCR® assay is a 10X assay designed to detect and quantify mutations in codons 12 and 13 of the KRAS gene using the ruby chip. KRAS is pivotal in regulating cell signaling pathways implicated in cancer development, notably melanoma and colorectal cancer.
  • Ready-to-use assay reagents to detect multiple targets in a single reaction
  • Crystal flex probe detection chemistry for flexible assay configuration
  • Same set of crystal universal reporters to detect any target in different assays
Wet lab tested
Compatibility: Nio™ Digital PCR, Prism6, Prism3, ruby chip
Channels: Blue, green
Required reagents: Crystal universal reporter 3 and naica® PCR MIX 10X
Targets: KRAS G12-G13
Caution: For research use only. Not for use in diagnostic procedures.