NANOCOLOR® Photometric Tube Tests, Chloride
Supplier: CTL SCIENTIFIC SUPPLY CORP
985019 985021
77578-444EA 133.34 CAD
77578-444 77578-446
NANOCOLOR® Photometric Tube Tests, Chloride
Environmental Test Kits
Pre-dosed NANOCOLOR® tube tests for photometric determination of chloride in water.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Barcode cuvette identification
  • Precisely pre-dosed reagents in 16 mm (Ext.Ø) tubes