NANOCOLOR® Photometric Tube Tests, COD
Supplier: CTL SCIENTIFIC SUPPLY CORP
963026
963029
985020
985022
985023
985026
985027
985028
985029
985030
985033
985036
985038
985011
985012
77578-464EA
87.97
CAD
77578-464
77578-458
77578-472
77578-474
77578-454
77578-462
77578-468
77578-460
77578-456
77578-476
77578-466
77578-482
77578-480
77578-470
77578-478
NANOCOLOR® Photometric Tube Tests, COD
Environmental Test Kits
A range of pre-dosed NANOCOLOR® tube tests for photometric determination of the COD (Chemical oxygen demand) in water.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Bar code cuvette identification
- Precisely pre-dosed reagents in 16 mm (Ext.Ø) tubes
The tests are particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.