NANOCOLOR® Photometric Tube Tests, Ethanol
Supplier: Macherey-Nagel
NANOCOLOR®
985838
77578-494EA 374.3 CAD
77578-494
NANOCOLOR® Photometric Tube Tests, Ethanol
Environmental Test Kits
A range of pre-dosed NANOCOLOR® tube tests for photometric determination of ethanol in water.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Barcode cuvette identification
  • Precisely pre-dosed reagents in 16 mm (ext.Ø) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.