NANOCOLOR® Photometric Test Kits, BOD₅
Supplier: CTL SCIENTIFIC SUPPLY CORP
985822 985825
77578-434EA 207.72 CAD
77578-434 77578-436
NANOCOLOR® Photometric Test Kits, BOD₅
Environmental Test Kits
A pre-dosed NANOCOLOR tube test for photometric determination of BOD₅ in water.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Bar code cuvette identification
  • Precisely pre-dosed reagents in 16 mm (ext.Ø) tubes
Tests that are particularly suitable for the NANOCOLOR® photometers but can be used with other photometers.