NANOCOLOR® Photometric Tube Tests, Aluminum
Supplier: Macherey-Nagel
NANOCOLOR®
985098
77578-414EA 142.25 CAD
77578-414
NANOCOLOR® Photometric Tube Tests, Aluminum
Environmental Test Kits
A pre-dosed NANOCOLOR tube test for photometric determination of aluminum in water.
  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Barcode cuvette identification
  • Precisely pre-dosed reagents in 16 mm (ext.Ø) tubes
The test is particularly suitable for NANOCOLOR® photometers but can be used with other photometers as well.