Transfectamine™ mRNA Transfection Reagent
Supplier: AAT BIOQUEST INC
60030
60031
77462-940EA
341.44
CAD
77462-940
77462-942
Transfectamine™ mRNA Transfection Reagent
Transfection Reagents
Transfectamine™ mRNA Transfection reagent is a powerful and versatile transfection reagent designed to introduce a higher amount of mRNA into eukaryotic cells, or more specifically, into animal cells.
- Consistently high transfection efficiency
- Higher viability of transfected cells
- Does not require special medium
Transfectamine™ mRNA Transfection reagent does not require special medium and is easier to use compared to most of the commercial transfection reagents.
Caution: For research use only.